missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCKL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17005-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a uridine kinase. Uridine kinases catalyze the phosphorylation of uridine to uridine monophosphate. This protein has been shown to bind to Epstein-Barr nuclear antigen 3 as well as natural killer lytic-associated molecule. Ubiquitination of this protein is enhanced by the presence of natural killer lytic-associated molecule. In addition, protein levels decrease in the presence of natural killer lytic-associated molecule, suggesting that association with natural killer lytic-associated molecule results in ubiquitination and subsequent degradation of this protein. Alternative splicing results in multiple transcript variants.Specifications
| UCKL1 | |
| Polyclonal | |
| Unconjugated | |
| UCKL1 | |
| UCK1 LIKE, UCK1L, UCKL1, Uridine cytidine kinase like 1, URKL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 499956, 54963, 68556 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91YL3, Q9NWZ5 | |
| UCKL1 | |
| UCKL1 Fusion Protein Ag10701 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction