missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCKL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118336
This item is not returnable.
View return policy
Description
UCKL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
Specifications
| UCKL1 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9NWZ5 | |
| Uckl1 | |
| E. coli-derived Human UCKL1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Uckl1 | |
| 1110007H10Rik; F538; RGD1565465; UCK1L; UCK1-LIKE; Uckl1; uridine kinase-like 1; uridine-cytidine kinase 1 like 1; uridine-cytidine kinase 1-like 1; uridine-cytidine kinase-like 1; URKL1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 54963 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction