missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCK2 Antibody (OTI21A6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44951-100ul
This item is not returnable.
View return policy
Description
UCK2 Monoclonal antibody specifically detects UCK2 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| UCK2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI21A6 | |
| Western Blot 1:500 | |
| Cytidine monophosphokinase 2, DKFZp686M04245, EC 2.7.1.48, Testis-specific protein TSA903, TSA903, UCK 2, UK, UMPK, uridine kinase, uridine monophosphate kinase, Uridine monophosphokinase 2, uridine-cytidine kinase 2 | |
| Synthetic peptide corresponding to residues near C-terminus of human UCK2 (NP_036606). | |
| 100 μL | |
| Signal Transduction | |
| 7371 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction