missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCK Antibody (OTI1A12), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74779MFV610
This item is not returnable.
View return policy
Description
UCK Monoclonal antibody specifically detects UCK in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| UCK | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI1A12 | |
| Flow Cytometry | |
| Cytidine monophosphokinase 1, EC 2.7.1.48, FLJ12255, Uridine monophosphokinase 1, uridine-cytidine kinase 1, URK1UCK 1 | |
| Full length human recombinant protein of human UCK1 (NP_113620) produced in HEK293T cell. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 83549 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction