missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCHL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | UCHL5 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16878663
|
Proteintech
11527-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16868663
|
Proteintech
11527-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCH-L5 (ubiquitin carboxyl-terminal hydrolase isozyme L5), also known as UCH37, is a 329 amino acid protein that functions to edit polyubiquinated protein substrates. Since UCH-L5 has the potential to rescue ubiquinated proteins, including oncogenic proteins, from proteasomal degradation, it is likely that deregulation of UCH-L5 may affect tumor growth. Through associations with Smad7, UHC-L5 can dramatically upregulate TGFbeta-dependent gene expression by de-ubiquinating and stabilizing TGFbeta RI. Also, since overexpression of UCH-L5 and other deubiquitinating enzymes has been observed in many cancer cell lines, inhibition of these proteins may be of some interest in designing therapies for cancer treatment. There are four isoforms of UCH-L5 that exist as a result of alternative splicing events.Specifications
| UCHL5 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q9WUP7, Q9Y5K5 | |
| 360853, 51377, 56207 | |
| UCHL5 Fusion Protein Ag2095 | |
| Primary | |
| -20°C | |
| UCHL5 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CGI 70, INO80R, Ubiquitin thioesterase L5, UCH L5, UCH37, UCHL5 | |
| UCHL5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title