missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCHL3 Recombinant Rabbit Monoclonal Antibody (001)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560321
This item is not returnable.
View return policy
Description
UCHL3 Recombinant Monoclonal Antibody for ELISA
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
Specifications
| UCHL3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P15374 | |
| UCHL3 | |
| Recombinant Human UCHL3 protein (Glu2-Ala230). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 001 | |
| Unconjugated | |
| UCHL3 | |
| RGD1561196; testicular tissue protein Li 221; ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase); ubiquitin carboxyl-terminal hydrolase isozyme L3; ubiquitin C-terminal hydrolase L3; Ubiquitin thioesterase L3; ubiquitin thiolesterase; UCHL3; UCH-L3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7347 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction