missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UCH37 Recombinant Rabbit Monoclonal Antibody (K01_2J79)

Product Code. 30830112
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830112 100 μL
30830273 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830112 Supplier Invitrogen™ Supplier No. MA563911
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

UCH37 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

UCH-L5 (ubiquitin carboxyl-terminal hydrolase isozyme L5), also known as UCH37, is a 329 amino acid protein that functions to edit polyubiquinated protein substrates. Since UCH-L5 has the potential to rescue ubiquinated proteins, including oncogenic proteins, from proteasomal degradation, it is likely that deregulation of UCH-L5 may affect tumor growth. Through associations with Smad7, UHC-L5 can dramatically upregulate TGFβ-dependent gene expression by de-ubiquinating and stabilizing TGFβ RI. Also, since overexpression of UCH-L5 and other deubiquitinating enzymes has been observed in many cancer cell lines, inhibition of these proteins may be of some interest in designing therapies for cancer treatment. There are four isoforms of UCH-L5 that exist as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCH37
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2J79
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene UCHL5
Gene Accession No. Q9WUP7, Q9Y5K5
Gene Alias 5830413B11Rik; AD-019; CGI-70; INO80 complex subunit R; INO80R; ubiquitin carboxyl-terminal esterase L5; ubiquitin carboxyl-terminal hydrolase isozyme L5; ubiquitin carboxyl-terminal hydrolase L5; ubiquitin C-terminal hydrolase 37; ubiquitin C-terminal hydrolase L5; ubiquitin C-terminal hydrolase UCH37; ubiquitin thioesterase L5; UCH37; Uchl5; UCH-L5
Gene Symbols UCHL5
Host Species Rabbit
Immunogen A synthetic peptide of human UCH37.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360853, 51377, 56207
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.