missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L5/UCH37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £470.00
Specifications
| Antigen | UCH-L5/UCH37 |
|---|---|
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18459901
|
Novus Biologicals
NBP1-85656-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257906
|
Novus Biologicals
NBP1-85656 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCH-L5/UCH37 Polyclonal specifically detects UCH-L5/UCH37 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| UCH-L5/UCH37 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51377 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CTHQDVHLGETLSEFKEFSQSFDAAMKGLALSNSDVIRQVHNSFARQQMFEFDTKT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 3.4.19.12, INO80 complex subunit R, INO80R, ubiquitin carboxyl-terminal esterase L5, ubiquitin carboxyl-terminal hydrolase isozyme L5, ubiquitin carboxyl-terminal hydrolase L5, Ubiquitin C-terminal hydrolase UCH37, Ubiquitin thioesterase L5, UCH37CGI-70, UCH-L5 | |
| UCHL5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title