missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UCH-L3 Polyclonal antibody specifically detects UCH-L3 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | UCH-L3 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human UCH-L3. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?