missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UCH-L3 Monoclonal antibody specifically detects UCH-L3 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse UCH-L3 (Uniprot#: Q9JKB1; Glu2-Ala230). |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?