missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L3 Antibody (204), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90680AF594
This item is not returnable.
View return policy
Description
UCH-L3 Monoclonal specifically detects UCH-L3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| UCH-L3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 7347 | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 204 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse UCH-L3 (Q9JKB1; Glu2-Ala230). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human UCHL3 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction