missing translation for 'onlineSavingsMsg'
Learn More

UCH-L1/PGP9.5 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB300-676B

Product Code. 18220540

  • £420.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

UCH-L1/PGP9.5 Polyclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UCH-L1/PGP9.5
Polyclonal
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated
P09936
UCHL1
A synthetic peptide made to a C-terminal portion of the human UCHL1 protein (between residues 200-220). [UniProt# P09936]
Affinity Purified
RUO
Primary
Human and mouse. Predicted to react with monkey, rat, pig, and horse based on 100% sequence homology.
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Biotin
PBS with 0.05% Sodium Azide
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Rabbit
25 kDa
0.1 mL
Neuronal Cell Markers, Neuroscience, Neurotransmission
7345
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only