missing translation for 'onlineSavingsMsg'
Learn More

UCB9608, MedChemExpress

Product Code. 16490947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490947 50 mg 50mg
16484658 10 mg 10mg
16457377 5 mg 5mg
16455496 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490947 Supplier Medchem Express Supplier No. HY112613

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UCB9608 is a potent, selective and orally active PI4KIIIβ inhibitor, with an IC50 of 11 nM, selective over PI3KC2 α, β, and γ lipid kinases. UCB9608 improves metabolic stability and exhibits excellent pharmacokinetic profile, acts as a potent immunosuppressive agent.

UCB9608 is a potent, selective and orally active PI4KIIIβ inhibitor, with an IC50 of 11 nM, selective over PI3KC2 α, β, and γ lipid kinases. UCB9608 improves metabolic stability and exhibits excellent pharmacokinetic profile, acts as a potent immunosuppressive agent.

Specifications

Chemical Name or Material UCB9608
CAS 1616413-96-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H26N8O2
Quantity 50 mg
Solubility Information DMSO : 65 mg/mL (158.36 mM; Need ultrasonic)
SMILES NC1=NC(N2CCN(C(NC3=CC=C(OC)C=C3C)=O)C[C@@H]2C)=C4C(N(C)N=C4)=N1
Molecular Weight (g/mol) 410.47
Formula Weight 410.47
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.