missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN10 Antibody (OTI1H5), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74762PEATT594
This item is not returnable.
View return policy
Description
UBXN10 Monoclonal specifically detects UBXN10 in Human samples. It is validated for Flow Cytometry.
Specifications
| UBXN10 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| UBXN10 | |
| Full length recombinant protein of human UBXN10 (NP_689589) produced in HEK293T cells. | |
| 0.1 mL | |
| 127733.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI1H5 | |
| Flow Cytometry | |
| FLJ25429, UBX domain containing 3, UBX domain protein 10, UBX domain-containing protein 10, UBX domain-containing protein 3, UBXD3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction