missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBXN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531809
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (91%), Rat (87%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ubiquitin-binding protein that plays a role in the modulation of innate immune response. Blocks both the RIG-I-like receptors (RLR) and NF-kappa-B pathways. Following viral infection, UBXN1 is induced and recruited to the RLR component MAVS. In turn, interferes with MAVS oligomerization, and disrupts the MAVS/TRAF3/TRAF6 signalosome. This function probably serves as a brake to prevent excessive RLR signaling (PubMed:23545497). Interferes with the TNFalpha-triggered NF-kappa-B pathway by interacting with cellular inhibitors of apoptosis proteins (cIAPs) and thereby inhibiting their recruitment to TNFR1 (PubMed:25681446). Prevents also the activation of NF-kappa-B by associating with CUL1 and thus inhibiting NF-kappa-B inhibitor alpha/NFKBIA degradation that remains bound to NF-kappa-B (PubMed:28152074). Interacts with the BRCA1-BARD1 heterodimer and regulates its activity. Specifically binds 'Lys-6'-linked polyubiquitin chains. Interaction with autoubiquitinated BRCA1 leads to the inhibition of the E3 ubiquitin-protein ligase activity of the BRCA1-BARD1 heterodimer (PubMed:20351172). Component of a complex required to couple deglycosylation and proteasome-mediated degradation of misfolded proteins in the endoplasmic reticulum that are retrotranslocated in the cytosol. [UniProt].
Specifications
| UBXN1 | |
| Polyclonal | |
| Unconjugated | |
| UBXN1 | |
| 2B28; 4930455J02Rik; D19Ertd721e; mY33K; Protein 2B28; RGD1309471; SAKS1; SAPK substrate protein 1; T25529; UBA/UBX 33.3 kDa protein; UBX domain protein 1; UBX domain-containing protein 1; UBXD10; UBXN1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 225896, 51035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.78 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q04323, Q922Y1 | |
| UBXN1 | |
| Recombinant protein fragment corresponding to a region within amino acids 106 and 297 of Human UBXN1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction