missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBXD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555914
This item is not returnable.
View return policy
Description
Immunogen sequence: SENGEQAFLL MMQPDNTIGD VRALLAQARV MDASAFEIFS TFPPTLYQDD TLTLQAAGLV PKAALLLRAR RAPKSSLKFS PGP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 70%.
This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3' coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| UBXD5 | |
| Polyclonal | |
| Unconjugated | |
| UBXN11 | |
| 4930506L07Rik; COA-1; colorectal tumor-associated antigen COA-1; colorectal tumor-associated antigen-1; D4Bwg1540e; PP2243; Soc; SOCI; Socius; UBX domain containing 5; UBX domain protein 11; UBX domain-containing protein 11; UBX domain-containing protein 5; UBXD5; Ubxn11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91544 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T124 | |
| UBXN11 | |
| Recombinant protein corresponding to Human UBXD5. Recombinant protein control fragment (Product #RP-94323). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction