missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | UBXD2 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629938
|
Novus Biologicals
NBP3-05991-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674988
|
Novus Biologicals
NBP3-05991-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBXD2 Polyclonal antibody specifically detects UBXD2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| UBXD2 | |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| erasin, FLJ23318, KIAA0242KIAA2042, UBX domain containing 2, UBX domain protein 4, UBX domain-containing 1, UBX domain-containing 2, UBX domain-containing protein 2, UBX domain-containing protein 4, UBXD2, UBXDC1 | |
| Produced in rabbits immunized with E. coli-derived Human UBXD2 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 23190 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title