missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBTD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103263
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total UBTD2.
UBTD2 gene ontology (GO) annotation includes cytoplasm and cytoplasm.
Specifications
| UBTD2 | |
| Polyclonal | |
| Unconjugated | |
| UBTD2 | |
| 4930571L24; 9630054F20Rik; AI645720; DCUBP; DC-UbP; Dendritic cell-derived ubiquitin-like protein; RGD1305506; SB72; ubiquitin domain containing 2; ubiquitin domain-containing protein 2; Ubiquitin-like protein SB72; Ubtd2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287178, 327900, 92181 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6PGH0, Q8WUN7 | |
| UBTD2 | |
| A synthesized peptide derived from human UBTD2(Accession Q8WUN7), corresponding to amino acid residues L181-P231. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction