missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBTD2, Mouse anti-Human, Clone: 1B8-1B1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00092181-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MTDGQLRSKRDEFWDTAPAFEGRKEIWDALKAAAHAFESNDHELAQAIIDGANITLPHGALTECYDELGNRYQLPVYCLAPPINMIEEKSDIETLDIPEPPPNSGYECQLRLRLSTGKDLKLVVRSTDTVFHMKRRLHAAEGVEPGSQRWFFSGRPLTDKMKFEELKIPKDYVVQVIVSQPVQNPTPVENSpecifications
| UBTD2 | |
| Monoclonal | |
| Unconjugated | |
| ubiquitin domain containing 2 | |
| DC-UbP/DCUBP/MGC30022 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1B8-1B1 | |
| In 1x PBS, pH 7.4 | |
| BC019910 | |
| UBTD2 | |
| DC-UbP (AAH19910, 1 a.a. ∼ 190 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 92181 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering