missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBR5/EDD Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71802R
This item is not returnable.
View return policy
Description
UBR5/EDD Polyclonal antibody specifically detects UBR5/EDD in Human, Mouse samples. It is validated for Western Blot
Specifications
| UBR5/EDD | |
| Polyclonal | |
| Western Blot | |
| DD5, E3 ubiquitin protein ligase, HECT domain containing, 1, E3 ubiquitin-protein ligase, HECT domain-containing 1, EC 6.3.2, EC 6.3.2.-, EDD1E3 ubiquitin-protein ligase UBR5, EDDE3 identified by differential display, hHYD, HYDFLJ11310, Hyperplastic discs protein homolog, KIAA0896MGC57263, progestin induced protein, Progestin-induced protein, ubiquitin protein ligase E3 component n-recognin 5, ubiquitin-protein ligase | |
| The immunogen for this product maps to a region between residue 2749 and 2799 of human E3 Ubiquitin Protein Ligase, HECT domain using the numbering given in entry NP_056986.2 (GeneID 51366). | |
| 0.1 mL | |
| DNA Repair | |
| 51366 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction