missing translation for 'onlineSavingsMsg'
Learn More

UBR4 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18411726 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411726 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411726 Supplier Novus Biologicals Supplier No. NBP128730AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBR4 Polyclonal antibody specifically detects UBR4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBR4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 600 kDa retinoblastoma protein-associated factor, EC 6.3.2, EC 6.3.2.-, KIAA0462E3 ubiquitin-protein ligase UBR4, KIAA1307zinc finger, UBR1 type 1, N-recognin-4, p600, RBAF600FLJ41863, Retinoblastoma-associated factor of 600 kDa, ubiquitin protein ligase E3 component n-recognin 4, Zinc finger UBR1-type protein 1, ZUBR1retinoblastoma-associated factor 600-like protein
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1350 and 1400 of human retinoblastoma-associated factor 600 using the numbering given in entry NP_065816.1 (GeneID 23352).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23352
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.