missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555715
This item is not returnable.
View return policy
Description
Immunogen sequence: QPNLTQWIRT ISQQIKALQF LRKEESTPNN ASTKNSENVD ELQLPEGFRP DFRPKIPYSE SIKEMLTTFG TATYKVGLKV HPN Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 65%.
The mammalian NUMB homolog plays a role in the determination of cell fate during development and binds with a variety of proteins, including Eps15, LNX1 and Notch 1. NumbL (NUMB-like protein), also known as Numb-R, NBL, CAG3A, CTG3a, NUMBLIKE or TNRC23, is a 609 amino acid cytoplasmic protein that, like NUMB, is thought to play a role in cell fate. Expressed at high levels in developing brain tissue, NumbL contains one PID (phosphotyrosine interaction domain) and plays an important role in neuronal differentiation, possibly associating with Eps15 and Notch 1. In mice, deletion of the NumbL gene is associated with early embryonic death, suggesting an essential role for NumbL in early development.
Specifications
| UBR2 | |
| Polyclonal | |
| Unconjugated | |
| UBR2 | |
| 9930021A08Rik; AI462103; AW540746; bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; E130209G04Rik; E3 ubiquitin-protein ligase UBR2; Kiaa0349; mKIAA0349; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; si:ch211-239i4.2; ubiquitin ligase E3 alpha-II; ubiquitin protein ligase E3 component n-recognin 2; ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II; Ubr2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23304 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IWV8 | |
| UBR2 | |
| Recombinant protein corresponding to Human UBR2. Recombinant protein control fragment (Product #RP-94602). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction