missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBR2, Mouse anti-Human, Clone: 4G4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00023304-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCGSpecifications
| UBR2 | |
| Monoclonal | |
| Unconjugated | |
| ubiquitin protein ligase E3 component n-recognin 2 | |
| C6orf133/DKFZp686C08114/KIAA0349/MGC71112/RP3-392M17.3/bA49A4.1/dJ242G1.1/dJ392M17.3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, KnockDown, Western Blot | |
| 4G4 | |
| In 1x PBS, pH 7.4 | |
| NM_015255 | |
| UBR2 | |
| UBR2 (NP_056070, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 23304 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction