missing translation for 'onlineSavingsMsg'
Learn More

UBPY/USP8 Antibody [FITC], Novus Biologicals™

Product Code. 30733432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733432 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733432 Supplier Novus Biologicals Supplier No. AF7735F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

UBPY/USP8 Polyclonal antibody specifically detects UBPY/USP8 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBPY/USP8
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias Deubiquitinating enzyme 8, EC 3.1.2.15, EC 3.4.19.12, HumORF8, KIAA0055hUBPy, MGC129718, ubiquitin carboxyl-terminal hydrolase 8, Ubiquitin isopeptidase Y, ubiquitin specific peptidase 8, ubiquitin specific protease 8, ubiquitin thioesterase 8, Ubiquitin thiolesterase 8, Ubiquitin-specific-processing protease 8, UBPYFLJ34456
Host Species Sheep
Immunogen E. coli-derived recombinant human USP8, Met1-Thr1118, Accession # P40818
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9101
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.