missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13856-1-AP-20UL
This item is not returnable.
View return policy
Description
UBP1 functions as a transcriptional activator in a promoter context-dependent manner. Modulates the placental expression of CYP11A1. Involved in regulation of the alpha-globin gene in erythroid cells. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with TFCP2. Involved in regulation of the alpha-globin gene in erythroid cells. Binds strongly to sequences around the HIV-1 initiation site and weakly over the TATA-box. Represses HIV-1 transcription by inhibiting the binding of TFIID to the TATA-box.Specifications
| UBP1 | |
| Polyclonal | |
| Unconjugated | |
| UBP1 | |
| LBP 1a, LBP 1B, LBP1, LBP1A, LBP1B, Transcription factor LBP 1, UBP1, Upstream binding protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22221, 301038, 7342 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q811S7, Q9NZI7 | |
| UBP1 | |
| UBP1 Fusion Protein Ag4830 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction