missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110678
This item is not returnable.
View return policy
Description
Immunogen sequence: TAYVNNSPSP APTFTSPQQS TCSVPDSNSS SPNHQGDGAS QTSGEQIQPS ATIQETQQWL LKNRFSSYTR LFSNFSGADL LKLTKEDLVQ ICGAADGIRL YNSLKSRSVR PRLTIYVCRE QPSSTVLQGQ QQAASSASEN GSGA Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 90%.
UBP1 functions as a transcriptional activator in a promoter context-dependent manner. Modulates the placental expression of CYP11A1. Involved in regulation of the alpha-globin gene in erythroid cells. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with TFCP2. Involved in regulation of the alpha-globin gene in erythroid cells. Binds strongly to sequences around the HIV-1 initiation site and weakly over the TATA-box. Represses HIV-1 transcription by inhibiting the binding of TFIID to the TATA-box.
Specifications
| UBP1 | |
| Polyclonal | |
| Unconjugated | |
| UBP1 | |
| Cp2b; LBP1; LBP1A; LBP-1a; LBP1B; LBP-1b; NF2d9; Nuclear factor 2d9; RGD1564700; transcription factor LBP-1; Ubp1; upstream binding protein 1; upstream binding protein 1 (LBP-1a); upstream-binding protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7342 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZI7 | |
| UBP1 | |
| Recombinant Protein corresonding to Human UBP1. Recombinant protein control fragment (Product #RP-102299). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction