missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ UBP 296

Product Code. 18054422
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18054422 10 mg 10mg
18035522 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054422 Supplier R&D Systems™ Supplier No. 2078/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective kainate antagonist; selective for GluK1-containing receptors

Selective GluK1 (formally GluR5) subunit containing kainate receptor antagonist (apparent KD = 1.09 μM). Displays ∽ 90-fold selectivity over AMPA receptors and recombinant hGluK2 (formally hGluR6) and GluK5 (formally KA2) containing kainate receptors. Has little or no action at NMDA or group I mGlu receptors. Selectively blocks kainate receptor-mediated LTP induction in rat hippocampal mossy fibers.

Active Enantiomer also available.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745055-86-1
Molecular Formula C15H15N3O6

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 333.3
Purity >98%
Molecular Weight (g/mol) 333.3
Chemical Name or Material UBP 296
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.