missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UBN1 Monoclonal antibody specifically detects UBN1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Applications | Western Blot |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HIRA-binding protein, ubinuclein 1, ubinuclein-1, VT, VT4 |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1. |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 29855 |
| Target Species | Human |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?