missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ubl5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570203
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 92%; Zebrafish: 86%.
This gene encodes a member of a group of proteins similar to ubiquitin. The encoded protein is not thought to degrade proteins like ubiquitin but to affect their function through being bound to target proteins by an isopeptide bond. The gene product has been studied as a link to predisposition to obesity based on its expression in Psammomys obesus, the fat sand rat, which is an animal model for obesity studies. Variation in this gene was found to be significantly associated with some metabolic traits (PMID: 15331561) but not associated with childhood obesity (PMID: 19189687). Pseudogenes of this gene are located on chromosomes 3, 5 and 17. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2013].
Specifications
| Ubl5 | |
| Polyclonal | |
| Unconjugated | |
| UBL5 | |
| 1110030M22Rik; beacon; HUB1; similar to ubiquitin-like 5; testicular tissue protein Li 217; ubiquitin like 5; ubiquitin-like 5; ubiquitin-like 5 protein; ubiquitin-like protein 5; Ubl5; Unknown (protein for MGC:127118); zgc:66388 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 500954, 59286 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BZL1 | |
| UBL5 | |
| synthetic peptide directed towards the middle region of human Ubl5 (aa 18-67). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction