missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBL5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-470
This item is not returnable.
View return policy
Description
UBL5 Polyclonal specifically detects UBL5 in Yeast samples. It is validated for Western Blot, ELISA.
Specifications
| UBL5 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| FLJ46917, HUB1, MGC131795, ubiquitin-like 5, ubiquitin-like protein 5 | |
| Rabbit | |
| 8.27 kDa | |
| 0.5 mg | |
| Neuroscience, Vision | |
| 59286 | |
| Reconstitute with 100 ul deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 1:500-1:2000, ELISA 1:1000-1:5000 | |
| Q6Q546 | |
| UBL5 | |
| This purified UBL5 Antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast UBL5 protein. (Uniprot: Q6Q546) | |
| Multi-step | |
| RUO | |
| Primary | |
| Hub 1 | |
| Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction