missing translation for 'onlineSavingsMsg'
Learn More

UBL4A Antibody (OTI1D4), DyLight 594, Novus Biologicals™

Product Code. 18421365 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421365 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421365 Supplier Novus Biologicals Supplier No. NBP274752DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBL4A Monoclonal antibody specifically detects UBL4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBL4A
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1D4
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DXS254EUbiquitin-like protein GDX, G6PD, GDXubiquitin-like 4, GET5, MDY2, TMA24, ubiquitin-like 4A, ubiquitin-like protein 4A, UBL4DX254E
Host Species Mouse
Immunogen Full length recombinant protein of human UBL4A (NP_055050) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 8266
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.