missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14100-1-AP-150UL
This item is not returnable.
View return policy
Description
Ubiquitin is a 77 amino acid protein that targets proteins for degradation by the 26S proteasome. Ubiquitin-like proteins are not directly involved in protein degradation, but appear to have many mechanistic similarities with the ubiquitin pathway. UBL3 (ubiquitin-like protein 3), also known as membrane-anchored ubiquitin-fold protein (MUB) or PNSC1, is a 117 amino acid membrane protein belonging to the ubiquitin-like family. Highly conserved between species, UBL3 contains two potential N-glycosylation sites, a potential protein kinase C phosphorylation site, and a potential C-terminal prenylation site. The gene encoding UBL3 is localized to chromosome 13q12.3.Specifications
| UBL3 | |
| Polyclonal | |
| Unconjugated | |
| UBL3 | |
| HCG 1, HsMUB, MUB, PNSC1, Protein HCG 1, ubiquitin like 3, Ubiquitin like protein 3, UBL3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24109, 363869, 5412 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95164, Q5BJT2, Q9Z2M6 | |
| UBL3 | |
| UBL3 Fusion Protein Ag5233 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction