Learn More
Ubiquitin Rabbit Polyclonal Antibody, Epredia™
Ubiquitin, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Brand: Epredia RB-9203-P
Description
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by a 26S complex, multicatalytic cytosolic and nuclear protease termed proteasome. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant full-length human ubiquitin protein
Molecular Weight: 8.5kDa
Positive Control: Human brain from patient with Alzheimer's disease
Cellular Localization: Nuclear, cytoplasmic and membrane This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Ubiquitin | |
| Polyclonal | |
| 200µg/mL | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Unconjugated | |
| Recombinant full-length human ubiquitin protein | |
| IVD | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.