missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ubiquitin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10201-2-AP-20UL
This item is not returnable.
View return policy
Description
Ubiquitin is a highly conserved globular 76 amino acid protein of about 8. 5 kDa molecular weight. It has a important role in the targeting of proteins for proteolytic degradation. Proteins to be degraded are covalently coupled to the C-terminus of ubiquitin by means of ubiquitin ligases. The ubiquitin itself is frequently also ubiquitinated, producing a polyubiquitin chain.Specifications
| ubiquitin | |
| Polyclonal | |
| Unconjugated | |
| UBB | |
| Polyubiquitin B, UBB, ubiquitin, ubiquitin B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192255, 22187, 7314 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P0CG47, P0CG49, P0CG51 | |
| UBB | |
| ubiquitin Fusion Protein Ag0260 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction