missing translation for 'onlineSavingsMsg'
Learn More

Ubiquitin Isopeptidase Inhibitor I, G5, MedChemExpress

Product Code. p-200100481
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475877

Brand: Medchem Express HY100738

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ubiquitin Isopeptidase Inhibitor I, G5 (NSC 144303) is an apoptosome-independent caspase and apoptosis activator with IC50 values of 1.76 and 1.6 μM in E1A and E1A/C9DN cells, respectively.

Ubiquitin Isopeptidase Inhibitor I, G5 (NSC 144303) is an apoptosome-independent caspase and apoptosis activator with IC50 values of 1.76 and 1.6 μM in E1A and E1A/C9DN cells, respectively.

Specifications

Chemical Name or Material Ubiquitin Isopeptidase Inhibitor I, G5
Physical Form Solid
CAS 108477-18-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H14N2O7S
For Use With (Application) Cancer-programmed cell death
Synonym NSC144303
Solubility Information DMSO : 50 mg/mL (120.66 mM; Need ultrasonic) ∣THF : 3.44 mg/mL (8.30 mM; Need ultrasonic) ∣Acetone : 2.86 mg/mL (6.90 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(/C(C1)=C/C2=CC=C([N+]([O-])=O)C=C2)/C(CS1(=O)=O)=C/C3=CC=C([N+]([O-])=O)C=C3
Molecular Weight (g/mol) 414.39
Formula Weight 414.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.