missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Isopeptidase Inhibitor I, G5, MedChemExpress
Description
Ubiquitin Isopeptidase Inhibitor I, G5 (NSC 144303) is an apoptosome-independent caspase and apoptosis activator with IC50 values of 1.76 and 1.6 μM in E1A and E1A/C9DN cells, respectively.
Specifications
Specifications
| Chemical Name or Material | Ubiquitin Isopeptidase Inhibitor I, G5 |
| Physical Form | Solid |
| CAS | 108477-18-5 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H14N2O7S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | NSC144303 |
| Solubility Information | DMSO : 50 mg/mL (120.66 mM; Need ultrasonic) ∣THF : 3.44 mg/mL (8.30 mM; Need ultrasonic) ∣Acetone : 2.86 mg/mL (6.90 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?