missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ubiquitin B Recombinant Rabbit Monoclonal Antibody (K01_2J78)

Product Code. 30830296
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30830296 50 μL 50µL
30830077 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830296 Supplier Invitrogen™ Supplier No. MA565369
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Ubiquitin B Recombinant Monoclonal Antibody for IHC (P), Western Blot

Ubiquitin is a highly conserved protein of about 8.5 kDa molecular weight, which has an ATP dependent role in the targeting of proteins for proteolytic degradation. To perform this function, the protein to be degraded is first covalently attached to the C terminus of ubiquitin, and the ubiquitinated complex is then recognized by a complex of degradative enzymes. Interestingly, ubiquitin also becomes covalently bonded to many types of pathological inclusions, which appear to be resistant to normal degradation. Therefore, ubiquitin antibodies are very useful for studies of these inclusions. For example, the neurofibrillary tangles and paired helical filaments diagnostic of Alzheimer's disease, Lewy bodies seen in Parkinson's disease, and Pick bodies found in Pick's disease are all heavily ubiquitinated and can be readily visualized with ubiquitin antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2J78
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene UBB
Gene Accession No. P0CG47
Gene Alias AL033289; epididymis secretory protein Li 50; HEL-S-50; HUBCEP52; im:6892314; im:6892314 protein; MGC127041; polyubiquitin; polyubiquitin B; polyubiquitin-B; RPL40; Rps27a; si:ch211-202a12.3; Uba52; Ubb; Ubb2; UBC; Ubi-p63E; Ubiquitin; ubiquitin B; ubiquitin C; Ubiquitin-related; zgc:172187; zUBC
Gene Symbols UBB
Host Species Rabbit
Immunogen A synthetic peptide of human Ubiquitin.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7314
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.