missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin B Mouse anti-Human, Clone: UBB/2122, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | Ubiquitin B |
|---|---|
| Clone | UBB/2122 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Description
Ubiquitin B Monoclonal specifically detects Ubiquitin B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Ubiquitin B | |
| 0.2 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
| Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| UBB/2122 | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 7314 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title