missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin B Antibody (UBB/2122), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08906F
This item is not returnable.
View return policy
Description
Ubiquitin B Monoclonal antibody specifically detects Ubiquitin B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Ubiquitin B | |
| Monoclonal | |
| FITC | |
| FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
| Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary) | |
| 100 μg | |
| Alzheimers Research, Apoptosis, Cell Biology, Neurodegeneration, Neuroscience | |
| 7314 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| UBB/2122 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction