missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Specifications
Specifications
| Gene Alias | RPS27A, UBA52, UBB ubiquitin B, UBC |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Gene ID (Entrez) | 6233 |
| Target Species | Human, Pig |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?