missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Antibody (P4G7-H11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-19306
This item is not returnable.
View return policy
Description
Ubiquitin Monoclonal specifically detects Ubiquitin in Human, Mouse, Rat, Bovine, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Ubiquitin | |
| Monoclonal | |
| Unconjugated | |
| PBS containing 50% glycerol with 0.09% Sodium Azide | |
| RPS27A | |
| Native bovine Ubiquitin | |
| 0.05 mg | |
| Alzheimers Research, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway | |
| 6233 | |
| Human, Mouse, Rat, Bovine, Hamster, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| P4G7-H11 | |
| Western Blot 1:100-1:2000, Flow Cytometry 10 ug/ml, Immunohistochemistry | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Ubiquitin (P4G7-H11) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction