missing translation for 'onlineSavingsMsg'
Learn More

Ubiquitin-activating Enzyme/UBE1 Rabbit anti-Human, Mouse, Rat, Clone: 6H2I9, Novus Biologicals™

Product Code. 18025335 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18025335 20 μg 20µL
18320927 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025335 Supplier Novus Biologicals Supplier No. NBP31679220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Ubiquitin-activating Enzyme/UBE1 Monoclonal antibody specifically detects Ubiquitin-activating Enzyme/UBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin-activating Enzyme/UBE1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6H2I9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias A1S9T, A1S9T and BN75 temperature sensitivity complementing, A1ST, AMCX1, GXP1, MGC4781, POC20, POC20 centriolar protein homolog, Protein A1S9, SMAX2, UBA1, ubiquitin-activating enzyme E1 homolog A, UBA1A, UBE1A1S9, UBE1X, Ubiquitin-activating enzyme E1, ubiquitin-activating enzyme E1 (A1S9T and BN75 temperature sensitivitycomplementing), ubiquitin-like modifier activating enzyme 1, ubiquitin-like modifier-activating enzyme 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 959-1058 of human Ubiquitin-activating Enzyme/UBE1 (P22314).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7317
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.