missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin-activating Enzyme/UBE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-472
This item is not returnable.
View return policy
Description
Ubiquitin-activating Enzyme/UBE1 Polyclonal specifically detects Ubiquitin-activating Enzyme/UBE1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ubiquitin-activating Enzyme/UBE1 | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, ELISA 1:2000-1:10000, Immunohistochemistry 2 mg/ml-20 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P22314 | |
| UBA1 | |
| Ubiquitin-activating Enzyme/UBE1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to full length Human Ubiquitin-activating Enzyme/UBE1. (Uniprot: P22314) | |
| Protein A purified | |
| RUO | |
| Primary | |
| UBE1 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| A1S9T, A1S9T and BN75 temperature sensitivity complementing, A1ST, AMCX1, GXP1, MGC4781, POC20, POC20 centriolar protein homolog, Protein A1S9, SMAX2, UBA1, ubiquitin-activating enzyme E1 homolog A, UBA1A, UBE1A1S9, UBE1X, Ubiquitin-activating enzyme E1, ubiquitin-activating enzyme E1 (A1S9T and BN75 temperature sensitivitycomplementing), ubiquitin-like modifier activating enzyme 1, ubiquitin-like modifier-activating enzyme 1 | |
| Rabbit | |
| 117.85 kDa | |
| 0.25 mg | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Ubiquitin Proteasome Pathway | |
| 7317 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction