missing translation for 'onlineSavingsMsg'
Learn More

Ubiquinol, TRC

Product Code. 30011557
Change view
Click to view available options
Quantity:
1 mg
25 mg
250 mg
Unit Size:
1mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011557 25 mg 25mg
30011341 250 mg 250mg
17937924 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011557 Supplier Toronto Research Chemicals Supplier No. TRCU70050025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-78-9
Molecular Formula C59 H92 O4
Molecular Weight (g/mol) 865.36
Synonym 1,4-Benzenediol, 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaen-1-yl]-5,6-dimethoxy-3-methyl-, 1,4-Benzenediol, 2-(3,7,11,15,19,23,27,31,35,39-decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-, (all-E)-, 1,4-Benzenediol, 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl]-5,6-dimethoxy-3-methyl- (9CI), Hydroquinone, 2-(3,7,11,15,19,23,27,31,35,39-decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl- (6CI,7CI,8CI), 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaen-1-yl]-5,6-dimethoxy-3-methyl-1,4-benzenediol, Coenzyme Q10, dihydro-, Coenzyme Q10-hydroquinone, Dihydrocoenzyme Q10, Kaneka QH, Reduced coenzyme Q10, Reduced ubiquinone Q10, Ubiquinol 50, Ubiquinol
IUPAC Name 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaenyl]-5,6-dimethoxy-3-methylbenzene-1,4-diol
SMILES COc1c(O)c(C)c(C\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CCC=C(C)C)c(O)c1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C59H92O4/c1-44(2)24-15-25-45(3)26-16-27-46(4)28-17-29-47(5)30-18-31-48(6)32-19-33-49(7)34-20-35-50(8)36-21-37-51(9)38-22-39-52(10)40-23-41-53(11)42-43-55-54(12)56(60)58(62-13)59(63-14)57(55)61/h24,26,28,30,32,34,36,38,40,42,60-61H,15-23,25,27,29,31,33,35,37,39,41,43H2,1-14H3/b45-26+,46-28+,47-30+,48-32+,49-34+,50-36+,51-38+,52-40+,53-42+
Formula Weight 864.6996
Chemical Name or Material Ubiquinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.