missing translation for 'onlineSavingsMsg'
Learn More

Ubiquilin 2 Antibody (6H9), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18433718 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433718 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433718 Supplier Novus Biologicals Supplier No. NBP225164MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ubiquilin 2 Monoclonal antibody specifically detects Ubiquilin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), KnockDown, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquilin 2
Applications Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Gene Knock-Out, KnockDown
Classification Monoclonal
Clone 6H9
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2
Host Species Mouse
Immunogen Human Ubiquilin 2 expressed in and purified from E. coli [UniProt# Q9UHD9]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 29978
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.