missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquilin 1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£133.90 - £349.17
Specifications
| Antigen | Ubiquilin 1 |
|---|---|
| Concentration | 0.26 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This antibody detects 70kD band of UBQLN1.
PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer′s and Parkinson′s disease.Specifications
| Ubiquilin 1 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| Q9JJP9, Q9UMX0 | |
| 114590, 29979 | |
| Ubiquilin 1 Fusion Protein Ag20252 | |
| Primary | |
| -20°C | |
| UBQLN1 |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DA41, DSK2, hPLIC 1, PLIC 1, PLIC1, ubiquilin 1, UBQLN1, XDRP1 | |
| Ubqln1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title