missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquilin 1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22126-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects 70kD band of UBQLN1.
PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer′s and Parkinson′s disease.Specifications
| Ubiquilin 1 | |
| Polyclonal | |
| Unconjugated | |
| UBQLN1 | |
| DA41, DSK2, hPLIC 1, PLIC 1, PLIC1, ubiquilin 1, UBQLN1, XDRP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114590, 29979 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JJP9, Q9UMX0 | |
| Ubqln1 | |
| Ubiquilin 1 Fusion Protein Ag17530 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction