missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubinuclein 1 Monoclonal Antibody (UBN1-02), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110458
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes the N-terminal portion of ubinuclein 1 (UBN1). Western blot results show a 165 kDa band.
Cellular senescence is a hallmark of tumor suppression and tissue aging. Senescent cells contain domains of heterochromatin, called senescence-associated heterochromatin foci (SAHF), that repress proliferation-promoting genes. The protein encoded by this gene binds to proliferation-promoting genes and is required for SAHF formation, enhancing methylation of histone H3.Specifications
Ubinuclein 1 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q9NPG3 | |
UBN1 | |
Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
UBN1-02 | |
Unconjugated | |
UBN1 | |
1110029L11Rik; 2610108L02Rik; AA673476; AW124741; HIRA-binding protein; Protein VT4; ubinuclein 1; ubinuclein-1; Ubiquitously expressed nuclear protein; Ubn1; VT; VT4 | |
Mouse | |
Protein A | |
RUO | |
29855 | |
4° C, do not freeze | |
Liquid |