missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBIAD1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA538035

Product Code. 15584961

  • £442.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A recommended positive control is mouse liver.

A novel gene designated TERE1, has a significant effect on the growth regulation in bladder cancer. The TERE1 gene maps to a chromosome locus that has been identified by loss of heterozygosity studies as a site of a putative tumor suppressor gene or genes for multiple tumor types including prostate carcinoma. Data suggest that TERE1 may be significant in prostate cancer growth regulation and the down regulation or absence of TERE1 may be an important component of the phenotype of advanced disease. Data also suggest a potential role for this gene product in the progression of bladder cancer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UBIAD1
Polyclonal
Unconjugated
UBIAD1
1200002M06Rik; AI426463; AW320947; RGD1309588; RP4-796F18.1; SCCD; Tere1; transitional epithelia response protein; transitional epithelial response protein 1; UbiA prenyltransferase domain containing 1; ubiA prenyltransferase domain-containing protein 1; UBIAD1
Rabbit
Antigen affinity chromatography
RUO
29914, 313706, 71707
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
D3ZG27, Q9DC60, Q9Y5Z9
UBIAD1
15 amino acid peptide near the carboxy terminus of human UBIAD1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.