missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBFD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565679
This item is not returnable.
View return policy
Description
Immunogen sequence: NKKEPLCRQKQ HRKVLDKGKP EDVMPSVKGA QERLPTVPLS GMYNKSGGKV RLTFKLEQDQ LWIGTKERTE KLPMGSIKNV VSEPIEGHED YHMMAFQLGP TEASYYWVYW VPTQ Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.
May play a role as NF-kappa-B regulator. [UniProt].
Specifications
| UBFD1 | |
| Polyclonal | |
| Unconjugated | |
| UBFD1 | |
| AI467302; D7Wsu105e; D7Wsu128e; RGD1306614; UBFD1; Ubiquitin domain-containing protein UBFD1; ubiquitin family domain containing 1; ubiquitin-binding protein homolog; UBPH | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56061 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14562 | |
| UBFD1 | |
| Recombinant Human UBFD1. Recombinant protein control fragment (Product #RP-106351). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction